missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dengue Virus NS5 Antibody - (methyltransferase domain), Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06447-50ul
This item is not returnable.
View return policy
Description
Dengue Virus NS5 Polyclonal antibody specifically detects Dengue Virus NS5 in Virus samples. It is validated for Western Blot, ELISA
Specifications
| Dengue Virus NS5 | |
| Polyclonal | |
| Western Blot 1:20000-1:50000, ELISA 1:5000-1:10000 | |
| Dengue NS5, Dengue NS5 protein | |
| Produced in rabbits immunized with purified, recombinant Dengue virus DENV-2 (Accession#: AAC59275.1; Gly2492-Gln2787). | |
| 50 μg | |
| Immunology, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites | |
| 1489713 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction