missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DENN/MADD Domain Containing 2D Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-30448
This item is not returnable.
View return policy
Description
DENN/MADD Domain Containing 2D Polyclonal specifically detects DENN/MADD Domain Containing 2D in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| DENN/MADD Domain Containing 2D | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Q9H6A0 | |
| DENND2D | |
| This antibody was developed against a recombinant protein corresponding to amino acids: REANGQGHFQERSFCKALTSKTNRRFVKKFVKTQLFSLFIQEAEKSKNPPAGYFQQKILEYEEQKKQKKPRE | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| DENN Domain-Containing Protein 2D, DENND2D, RP5-1180E21.2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 79961 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction