missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DENND4C Antibody (3E8), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11040C
This item is not returnable.
View return policy
Description
DENND4C Monoclonal antibody specifically detects DENND4C in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
Specifications
| DENND4C | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunofluorescence | |
| 300000000 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| bA513M16.3, C9orf55B, DENN/MADD domain containing 4C, DKFZp686I09113, FLJ20686 | |
| Full Length Human Recombinant DENND4C protein (NM_017925) | |
| 0.1 mL | |
| Alzheimers Research, Cellular Signaling, Cytokines and Growth Factors, Membrane Trafficking and Chaperones, Neurodegeneration, Neuroscience | |
| 55667 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction