Learn More
Invitrogen™ DENND4C Monoclonal Antibody (3E8), PerCP
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA545962
Product Code. 17847932
  • € 478.00 / 100µg
Description
A 1:1,000 dilution of MA5-45962 was sufficient for detection of DENND4C in A549, MEF cells by Western Blot analysis. Detects approximately 213 kDa.
DENND4C is guanine nucleotide exchange factor (GEF) activating RAB10. It promotes the exchange of GDP to GTP, converting inactive GDP-bound RAB10 into its active GTP-bound form. Thereby, stimulates SLC2A4/GLUT4 glucose transporter-enriched vesicles delivery to the plasma membrane in response to insulin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DENND4C
Monoclonal
1 mg/mL
2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4
A6H8H2, Q5VZ89
DENND4C
Full Length Human Recombinant Dennd4c protein.
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG1
Immunoprecipitation, Western Blot, Immunocytochemistry
3E8
PerCP
DENND4C
1700065A05Rik; AA420392; bA513M16.3; C9orf55; C9orf55B; DENN domain containing 4C; DENN domain-containing protein 4C; DENN domain-containing protein 4C-like protein; DENN/MADD domain containing 4A pseudogene; DENN/MADD domain containing 4C; Dennd4c; likely ortholog of H. sapiens chromosome 9 open reading frame 55 (C9orf55); RAB10GEF; RGD1308489
Mouse
Protein G
RUO
313340, 329877, 55667
4°C