missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DENND5B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 275.00 - € 547.00
Specifications
| Antigen | DENND5B |
|---|---|
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18482722
|
Novus Biologicals
NBP2-31971-25ul |
25 μL |
€ 275.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18112474
|
Novus Biologicals
NBP2-31971 |
0.1 mL |
€ 547.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DENND5B Polyclonal specifically detects DENND5B in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| DENND5B | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse | |
| Q6ZUT9 | |
| 160518 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: FIWDFIEKVVAYFETTDQILDNEDDVLIQKSSCKTFCHYVNAINTAPRNIGK | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| DENN domain-containing protein 5B, DENN/MADD domain containing 5B, DKFZp686P1174, FLJ41648, FLJ43333, MGC24039, Rab6IP1-like protein | |
| DENND5B | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title