missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DENR Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-50042AF350
This item is not returnable.
View return policy
Description
DENR Polyclonal antibody specifically detects DENR in Human, Mouse samples. It is validated for Western Blot
Specifications
| DENR | |
| Polyclonal | |
| Western Blot | |
| density-regulated protein, DRP1Protein DRP1, DRPsmooth muscle cell associated protein-3, SMAP-3, Smooth muscle cell-associated protein 3 | |
| The immunogen for this product maps to a region between residue 148 and 198 of human Density-Regulated Protein using the numbering given in entry NP_003668.2 (GeneID 8562). | |
| 0.1 mL | |
| Signal Transduction | |
| 8562 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction