Learn More
Thermo Scientific™ Dense Storage Racks for Chest ULTs and Cryo Storage
Shop All Thermo Scientific Products
Click to view available options
Holds:
Plates (10/Rack)
Plates (11/Rack)
Plates (12/Rack)
Plates (15/Rack)
Plates (18/Rack)
Plates (9/Rack)
Unit Size:
Each
Description
A convenient, streamlined process to maximize sample storage in your laboratory. Thermo Scientific™ Dense Storage Racks for Chest ULTs and Cryo Storage are designed to hold tubes ranging from 200μL up to 5.0μL, offering maximized sample storage capacity for ultra-low chest freezers and large-capacity cryogenic freezers.
- Warranty varies by country, please contact us for details
Specifications
Specifications
| Product Type | Tube Rack (Cryogenic and Ultra-Low Chest Freezers) |
| Description | Racks for Tubes (cryogenic and ultra-low chest freezers) |
| Color | Silver |
| Holds | Plates (9/Rack) |
| Shape | Rectangular |
| For Use With (Equipment) | Matrix 1 mL Screw-top Tubes |
| Freezer Compatibility (Capacity) | Thermo Scientific CryoPlus 1 (hold 7 racks), CryoPlus 2 (holds 16 racks), CryoPlus 3 (holds 30 racks), CryoPlus 4 (holds 51 racks) Thermo Scientific mechanical cryogenic chest freezer 6.8 cu. ft. (holds 18 racks), 10.3 cu. ft. (holds 27 racks) |
| Width (Metric) | 13.5 cm |
| Width (English) | 5.3 in. |
| Height (Metric) | 57.2 cm |
| Show More |