Learn More
Amarell™ Fine-Graduation Hydrometer without Thermometer, series M100
Product Code. 10272012
Click to view available options
Specific Gravity Range:
0.800 to 0.900 g/cm3
0.900 to1.000 g/cm3
1.000 to 1.100 g/cm3
1.100 to 1.200 g/cm3
Unit Size:
Each
Product Code. 10272012
Brand: Amarell™ H809608
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Calibrated for reference temperature 20°C. The high-graduation density hydrometer without thermometer is DIN rated for many specific gravity measurements.
  • Reference temperature 20°C
  • Each hydrometer length is 250mm
Specifications
Quantity 1Each
Type Density Hydrometer
Accuracy ±1g/cm3
Increments 0.0020 g/cm3
Length (Metric) 250 mm
Specific Gravity Range 1.000 to 1.100 g/cm3
Temperature of Standardization 20°C