Learn More
Amarell™ Fine-Graduation Hydrometer without Thermometer, series M100
Calibrated for reference temperature 20°C. The high-graduation density hydrometer without thermometer is DIN rated for many specific gravity measurements.
Brand:  Amarell™ H809610
Additional Details : Weight : 0.05000kg
 View more versions of this product
Product Code. 10282012
  • € 43.00 / Each
Estimated Shipment: 17-05-2024
to see stock.
Add to basket
Description
Description
  • Reference temperature 20°C
  • Each hydrometer length is 250mm
Specifications
Specifications
1Each
±1g/cm3
250 mm
20°C
Density Hydrometer
0.0020 g/cm3
1.100 to 1.200 g/cm3