Learn More
Density Standard (25°C); UKAS ISO/IEC17025 and ISO 17034 certified, nominal density 1.0242 g/mL (25°C), 500 mL, Paragon Scientific™
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Description
- Dual certified to ISO 17025 / ISO 17034 under our UKAS accreditation
- Certified in accordance with primary level ASTM D1480 methodology
- Ensures full compliance to ASTM and IP test method protocols
- Supplied in tamper evident packaging with a 12 month shelf life
- Manufactured in the United Kingdom
Specifications
Specifications
| Quantity | 500 mL |
Nominal values quoted. Certified Values will vary from batch to batch.