Learn More
Density Standard (40°C); UKAS ISO/IEC17025 and ISO 17034 certified, nominal density 0.8247 g/mL (40°C), 500 mL, Paragon Scientific™
Product Code. 16060222
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Product Code. 16060222
Brand: Paragon Scientific DEN4004500ML
Paragon ISO 17025 / ISO 17034 certified Density standards are for the verification of instruments used to measure density of materials which are fluid at the desired test temperature within the range of 15°C to 150°C
  • Dual certified to ISO 17025 / ISO 17034 under our UKAS accreditation
  • Certified in accordance with primary level ASTM D1480 methodology
  • Ensures full compliance to ASTM and IP test method protocols
  • Supplied in tamper evident packaging with a 2 year shelf life
  • Manufactured in the United Kingdom
Specifications
Quantity 500 mL
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Distillates (petroleum), hydrotreated light
Nominal values quoted. Certified Values will vary from batch to batch.