missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Density Standard Quality Range 0.7411 g/mL at 15°C, Reagecon
Shop All Reagecon Products

Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Description
- Density reference standard, density reference material.
- Tested by vibrational densitometer and can be used as a calibration or control standard for density measurement by vibrational or hydrometer (specific gravity) based techniques.
- Reagecon holds ISO 17025 Accreditation for testing density standards using vibrational densitometers (INAB Ref: 264T).
- The product is prepared gravimetrically on a weight/weight basis.
- Both solute and solvent are weighed on a balance calibrated by OIML traceable weights.
- Reagecon holds ISO 17025 Accreditation for calibration of laboratory balances (INAB Ref: 265C).
- The resulting Balance Certificate of Calibration is issued in accordance with the requirements of ISO/IEC 17025.
- The density of the product is established and certified using high performance calibrated vibrational densitometers.
- High accuracy (± 0.001642 g/mL).
- Tested using an ISO 17025 accredited test method.
Specifications
Specifications
| Packaging | Amber glass Winchester bottle |
| Physical Form | Liquid |
| Concentration | 0.7411 g/mL at 15°C (±0.001642) |
| For Use With (Equipment) | Vibrational Density Measuring Instrument |
| Quantity | 100 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction