missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Deoxyribonuclease II (Porcine)

Product Code. 16191820
Click to view available options
Quantity:
20,000 U
Unit Size:
20000 units
This item is not returnable. View return policy

Product Code. 16191820

Brand: Abnova™ P5297.20000Units

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for Enzyme

#REF!

Specifications

For Use With (Application) Enzyme Activity
Molecular Weight (g/mol) 38kDa
Name Deoxyribonuclease II (Porcine)
Preparation Method Native protein chromatographically purified from Porcine Spleen by a modification of the procedure of Bernardi et al., Biochim. Biophys. Acta, 129, 1 (1966)
Quantity 20,000 U
Storage Requirements Store at -20°C.
Regulatory Status RUO
Common Name DEOXYRIBONUCLEASE
Assays Deoxyribonuclease II Assay
Biological Activity ≥12,000 units per mg protein. One Unit causes an increase in absorbance at 260 nm of 0.001 per minute at 25°C, pH 4.6 using highly polymerized DNA as substrate.
Species Pig
Protein Tag None
Expression System Native protein chromatographically purified from Porcine Spleen by a modification of the procedure of Bernardi et al., Biochim. Biophys. Acta, 129, 1 (1966)
Form Lyophilized
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.