missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
DEP-1/CD148 Monoclonal antibody specifically detects DEP-1/CD148 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, CyTOF
Specifications
Specifications
| Antigen | DEP-1/CD148 |
| Applications | Western Blot, Flow Cytometry, Immunoprecipitation, CyTOF |
| Classification | Monoclonal |
| Clone | 143-41 |
| Conjugate | DyLight 405 |
| Dilution | Western Blot, Flow Cytometry, Immunoprecipitation, CyTOF-ready |
| Formulation | 50mM Sodium Borate |
| Gene Alias | CD148, CD148 antigen, Density-enhanced phosphatase 1, DEP-1, DEP1EC 3.1.3.48, HPTP eta, HPTPeta, human density enhanced phosphatase-1, protein tyrosine phosphatase, receptor type, J, protein tyrosine phosphatase, receptor type, J polypeptide, Protein-tyrosine phosphatase eta, Protein-tyrosine phosphatase receptor type J, receptor-type tyrosine-protein phosphatase eta, R-PTP-eta, R-PTP-J, SCC1, susceptibility to colon cancer 1, mouse, homolog of |
| Host Species | Mouse |
| Immunogen | Phytohemagglutinin-stimulated human peripheral blood mononuclear cells |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?