missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DEP-1/CD148 Antibody (261922), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB19341CL1
This item is not returnable.
View return policy
Description
DEP-1/CD148 Monoclonal antibody specifically detects DEP-1/CD148 in Human samples. It is validated for Flow Cytometry,CyTOF
Specifications
| DEP-1/CD148 | |
| Monoclonal | |
| CoraFluor 1 | |
| CD148, CD148 antigen, Density-enhanced phosphatase 1, DEP-1, DEP1EC 3.1.3.48, HPTP eta, HPTPeta, human density enhanced phosphatase-1, protein tyrosine phosphatase, receptor type, J, protein tyrosine phosphatase, receptor type, J polypeptide, Protein-tyrosine phosphatase eta, Protein-tyrosine phosphatase receptor type J, receptor-type tyrosine-protein phosphatase eta, R-PTP-eta, R-PTP-J, SCC1, susceptibility to colon cancer 1, mouse, homolog of | |
| E. coli-derived recombinant human DEP-1/CD148, Arg997-Ala1337, Accession # Q12913 | |
| 0.1 mL | |
| Protein Phosphatase | |
| 5795 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
| Flow Cytometry, CyTOF | |
| 261922 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction