Learn More
Invitrogen™ Depatuxizumab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA559384
 View more versions of this product
Product Code. 30283759
  • € 1262.00 / 1mg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Depatuxizumab has been used in trials studying the treatment and diagnostic of Adenocarcinoma and Advanced Solid Tumors.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Depatuxizumab Chimeric
Recombinant Monoclonal
Unconjugated
ABT-806
Protein A
RUO
Human
Antibody
IgG1
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Human
1 mg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized