missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DEPDC7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32141
This item is not returnable.
View return policy
Description
DEPDC7 Polyclonal specifically detects DEPDC7 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| DEPDC7 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| DEP domain containing 7, DEP domain-containing protein 7, dJ85M6.4, dJ85M6.4 (novel 58.3 KDA protein), novel 58.3 KDA protein, Protein TR2/D15, TR2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 91614 | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 1:1000 - 1:5000, Immunoprecipitation 2 - 10 mcg/mg lysate | |
| NP_001070710.1 | |
| DEPDC7 | |
| The immunogen this antibody was made to, maps to a region between residue 461 to 511 of human DEP Domain Containing 7 using the numbering given in entry NP_001070710.1 (GeneID 91614). | |
| 100 μL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction