missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DEPDC7 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32141PE
This item is not returnable.
View return policy
Description
DEPDC7 Polyclonal antibody specifically detects DEPDC7 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| DEPDC7 | |
| Polyclonal | |
| Western Blot, Immunoprecipitation | |
| DEP domain containing 7, DEP domain-containing protein 7, dJ85M6.4, dJ85M6.4 (novel 58.3 KDA protein), novel 58.3 KDA protein, Protein TR2/D15, TR2 | |
| The immunogen this antibody was made to, maps to a region between residue 461 to 511 of human DEP Domain Containing 7 using the numbering given in entry NP_001070710.1 (GeneID 91614). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| PE | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 91614 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction