missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DEPDC7, Mouse anti-Human, Clone: 4D9, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00091614-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: NPSEFKLQKESDNRMVVKRIFSKAIVDNKNLSKGKTDLLVLFLMDHQKDVFKIPGTLHKIVSVKLMAIQNGRDPNRDAGYIYCQRIDQRDYSNNTEKTTKDELLNLLSpecifications
| DEPDC7 | |
| Monoclonal | |
| Unconjugated | |
| DEP domain containing 7 | |
| TR2/dJ85M6.4 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunoprecipitation | |
| 4D9 | |
| In 1x PBS, pH 7.4 | |
| NM_139160 | |
| DEPDC7 | |
| LOC91614 (NP_631899, 393 a.a. ∼ 499 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 91614 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction