missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Depemokimab Humanized Recombinant Human Monoclonal Antibody

Product Code. 30132302
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132302 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30132302 Supplier Invitrogen™ Supplier No. MA554872

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Recombinant Monoclonal Antibody

Compatible with Direct ELISA.

Depemokimab is an anti-IL-5 monoclonal antibody that works by binding to interleukin-5 (IL-5), thereby inhibiting its interaction with the IL-5 receptor on the surface of eosinophils, leading to reduced eosinophil activation and survival.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Depemokimab Humanized
Applications ELISA
Classification Recombinant Monoclonal
Concentration 1.16 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias IL5
Host Species Human
Immunogen Recombinant Human IL5.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.