missing translation for 'onlineSavingsMsg'
Learn More

DEPP Rabbit anti-Human, Polyclonal, Proteintech

Product Code. p-7211434 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16824395 20 μL 20µL
16814395 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16824395 Supplier Proteintech Supplier No. 258331AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DEPP, also named as FIG and C10orf10, is known to be differentially regulated in mouse tissues in response to hypoxia and oxidative stress.

Specifications

Antigen DEPP
Applications Western Blot
Classification Polyclonal
Concentration 0.34 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene DEPP1
Gene Accession No. Q9NTK1
Gene Alias C10orf10, DEPP, Fasting induced gene protein, FIG, Protein DEPP
Gene Symbols DEPP1
Host Species Rabbit
Immunogen DEPP Fusion Protein Ag22773
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11067
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.