missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DERA Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565986
This item is not returnable.
View return policy
Description
Immunogen sequence: ISKIQVNHPAV LRRAEQIQAR RTVKKEWQAA WLLKAVTFID LTTLSGDDTS SNIQRLCYKA KYPIREDLLK ALNMHDKGIT TAAVCVYPAR VCDAVKALKA AGCNIPVAS Highest antigen sequence identity to the following orthologs - mouse 92%, rat 91%.
Catalyzes a reversible aldol reaction between acetaldehyde and D-glyceraldehyde 3-phosphate to generate 2-deoxy-D-ribose 5-phosphate. Participates in stress granule (SG) assembly. May allow ATP production from extracellular deoxyinosine in conditions of energy deprivation. [UniProt].
Specifications
| DERA | |
| Polyclonal | |
| Unconjugated | |
| DERA | |
| 2-deoxy-D-ribose 5-phosphate aldolase; 2-deoxyribose-5-phosphate aldolase homolog; CGI-26; DEOC; deoxyriboaldolase; Deoxyribose-phosphate aldolase; deoxyribose-phosphate aldolase (putative); DERA; Phosphodeoxyriboaldolase; putative deoxyribose-phosphate aldolase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51071 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y315 | |
| DERA | |
| Recombinant Human DERA. Recombinant protein control fragment (Product #RP-105030). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction