Learn More
Derazantinib, MedChemExpress
Product Code. 16477776
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16477776 100 mg 100mg
16467998 10 mg 10mg
16453726 25 mg 25mg
16433927 50 mg 50mg
16430336 2 mg 2mg
16400468 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 16477776 Supplier Medchem Express Supplier No. HY19981
MedChemExpress Derazantinib (ARQ-087) is an orally bioavailable, ATP competitive tyrosine kinase inhibitor; exhibits potent activity against FGFR1-3 chondrocytes with IC50s of 4.5, 1.8, and 4.5 nM, respectively.
Derazantinib (ARQ-087) is an orally bioavailable, ATP competitive tyrosine kinase inhibitor
Specifications
Chemical Name or Material Derazantinib
CAS 1234356-69-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C29H29FN4O
Quantity 100 mg
Synonym ARQ-087
Solubility Information DMSO : 25 mg/mL (53.35 mM; Need ultrasonic)
SMILES COCCNCCC1=CC(NC2=NC=C3C[C@@H](C4=CC=CC=C4F)C5=CC=CC=C5C3=N2)=CC=C1
Molecular Weight (g/mol) 468.57
Formula Weight 468.57
Percent Purity 98.68%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less
Research purposes only