missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DERL2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17267873 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17267873 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17267873 Supplier Invitrogen™ Supplier No. PA5115111

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total DERL2.

Functional component of endoplasmic reticulum-associated degradation (ERAD) for misfolded lumenal glycoproteins, but not that of misfolded nonglycoproteins. May act by forming a channel that allows the retrotranslocation of misfolded glycoproteins into the cytosol where they are ubiquitinated and degraded by the proteasome. May mediate the interaction between VCP and the degradation substrate. In contrast to DERL1, it is not involved in the degradation of MHC class I heavy chains following infection by cytomegaloviruses. May play a role in cell proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DERL2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene DERL2
Gene Accession No. Q8BNI4, Q9GZP9
Gene Alias carcinoma related; CGI-101; Degradation in endoplasmic reticulum protein 2; Der1-like domain family, member 2; der1-like protein 2; Der2; Derl2; derlin 2; derlin-2; derlin-2-like; DERtrin-2; F-LAN-1; Flana; F-LANa; LOC100910823; SBBI53
Gene Symbols DERL2
Host Species Rabbit
Immunogen A synthesized peptide derived from human DERL2(Accession Q9GZP9), corresponding to amino acid residues P193-G239.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100910823, 116891, 51009
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.