missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Derlin 1 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-448B
This item is not returnable.
View return policy
Description
Derlin 1 Polyclonal specifically detects Derlin 1 in Human, Mouse, Canine, Chinese Hamster samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Derlin 1 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| Q9BUN8 | |
| DERL1 | |
| A synthetic peptide made to the C-terminal region of human Derlin 1 (between residues 200-251). [UniProt# Q9BUN8] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Canine and Chinese Hamster. Predicted to react with Xenopus, bovine, chicken, rat, zebrafish, and mouse based on 100% sequence homology. Mouse reactivity reported in scientific literature (PMID: 25377857). | |
| Store at 4C in the dark. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| DER-1, DER1Degradation in endoplasmic reticulum protein 1, Der1-like domain family, member 1, Der1-like protein 1, derlin-1, DERtrin-1, FLJ13784, FLJ42092, MGC3067, PRO2577 | |
| Rabbit | |
| 29 kDa | |
| 0.1 mL | |
| ER Markers, Immunology, Protein Turnover, Signal Transduction, Unfolded Protein Response | |
| 79139 | |
| Human, Mouse, Canine, Chinese Hamster | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction