missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Derlin-2 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98598H
This item is not returnable.
View return policy
Description
Derlin-2 Polyclonal specifically detects Derlin-2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Derlin-2 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| carcinoma related, Degradation in endoplasmic reticulum protein 2, Der1-like domain family, member 2, Der1-like protein 2, DER2, derlin-2, F-LAN-1DERtrin-2, FLANa, F-LANaCGI-101 | |
| Produced in rabbits immunized with E. coli-derived Human Derlin-2 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| HRP | |
| PBS | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 51009 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction