missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DERP1 Antibody (06) [PerCP], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-33163PCP
This item is not returnable.
View return policy
Description
DERP1 Monoclonal antibody specifically detects DERP1 in Insect samples. It is validated for ELISA
Specifications
| DERP1 | |
| Monoclonal | |
| PerCP | |
| Der p 1 allergen, DERP-1 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant DERP1. | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
| ELISA | |
| 06 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Insect | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction