missing translation for 'onlineSavingsMsg'
Learn More

DERP1 Antibody (068), DyLight 680, Novus Biologicals™

Product Code. 30238968 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30238968 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30238968 Supplier Novus Biologicals Supplier No. NBP340580FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

DERP1 Monoclonal antibody specifically detects DERP1 in Insect samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen DERP1
Applications ELISA
Classification Monoclonal
Clone 068
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias Der p 1 allergen, DERP-1
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Dermatophagoides pteronyssinus DERP1. HEK293 used as expression system.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Insect
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.