missing translation for 'onlineSavingsMsg'
Learn More

Deruxtecan, MedChemExpress

Product Code. p-200098402
Encompass
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
This item is not returnable. View return policy

Product Code. 16431369

Brand: Medchem Express HY13631E

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Deruxtecan is an ADC drug-linker conjugate composed of a derivative of DX-8951 (DXd) and a maleimide-GGFG peptide linker, used for synthesizing DS-8201 and U3-1402.

Deruxtecan is an ADC drug-linker conjugate composed of a derivative of DX-8951 (DXd) and a

Specifications

Chemical Name or Material Deruxtecan
CAS 1599440-13-7
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C52H56FN9O13
Quantity 1 mg
Solubility Information DMSO : 35 mg/mL (33.85 mM; Need ultrasonic)
SMILES O=C(C=CC1=O)N1CCCCCC(NCC(NCC(N[C@@H](CC2=CC=CC=C2)C(NCC(NCOCC(N[C@@H]3C4=C5C(C(N6C5)=CC([C@](O)(C(OC7)=O)CC)=C7C6=O)=NC8=CC(F)=C(C)C(CC3)=C48)=O)=O)=O)=O)=O)=O
Molecular Weight (g/mol) 1034.05
Formula Weight 1034.05
Percent Purity 95.05%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.