Learn More
Des(methylsulfonyl) Dronedarone Oxalate, TRC
Specifications
Specifications
| Chemical Name or Material | Des(methylsulfonyl) Dronedarone Oxalate |
| CAS | 500791-70-8 |
| Molecular Formula | C34H46N2O11 |
| Quantity | 10 g |
| Synonym | (5-Amino-2-butyl-3-benzofuranyl)[4-[3-(dibutylamino)propoxy]phenyl]methanone Ethanedioate, Dronedarone USP Related Compound B |
| InChI Formula | InChI=1S/C30H42N2O3.2C2H2O4/c1-4-7-11-28-29(26-22-24(31)14-17-27(26)35-28)30(33)23-12-15-25(16-13-23)34-21-10-20-32(18-8-5-2)19-9-6-3;2*3-1(4)2(5)6/h12-17,22H,4-11,18-21,31H2,1-3H3;2*(H,3,4)(H,5,6) |
| SMILES | CCCCN(CCCC)CCCOc1ccc(cc1)C(=O)c2c(CCCC)oc3ccc(N)cc23.OC(=O)C(=O)O.OC(=O)C(=O)O |
| IUPAC Name | (5-amino-2-butyl-1-benzofuran-3-yl)-[4-[3-(dibutylamino)propoxy]phenyl]methanone;oxalic acid |
| Molecular Weight (g/mol) | 658.74 |
| Formula Weight | 658.3102 |