Learn More
Poulten & Graf GmbH Desalination Unit LAB-IoN L 2
Product Code. 10478611
Click to view available options
Includes:
4 diodes
8 diodes
Unit Size:
Each
Product Code. 10478611
Brand: Poulten & Graf GmbH 110 320
Only null left
Add to basket
Add to basket
With the desalination unit, Poulten & Graf GmbH Desalination Unit LAB-IoN L 2, demineralised water can be produced in the simplest way.
  • Used cartridges are simply replaced
  • Connection tube to a 3/4in. tap
  • Includes pH meter
  • Includes mounting
Specifications
Capacity (Metric) 30 to 40 L/hr.
Includes 4 diodes
Type Desalination Unit