missing translation for 'onlineSavingsMsg'
Learn More

Desmethyl Fluvoxamine, TRC

Product Code. 30135620
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135620 50 mg 50mg
17985115 5 mg 5mg
17930675 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135620 Supplier Toronto Research Chemicals Supplier No. TRCD29163050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 192876-02-1
Molecular Formula C14 H19 F3 N2 O2
Molecular Weight (g/mol) 304.31
Synonym (1E)-5-hydroxy-1-[4-(trifluoromethyl)phenyl]-1-Pentanone O-(2-aminoethyl)oxime, 1-Pentanone, 5-hydroxy-1-[4-(trifluoromethyl)phenyl]-, O-(2-aminoethyl)oxime, (E)-, Fluvoxamino acid, Fluvoxamine Maleate Imp. G (EP)
IUPAC Name (5E)-5-(2-aminoethoxyimino)-5-[4-(trifluoromethyl)phenyl]pentan-1-ol
SMILES NCCO\N=C(/CCCCO)\c1ccc(cc1)C(F)(F)F

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C14H19F3N2O2/c15-14(16,17)12-6-4-11(5-7-12)13(3-1-2-9-20)19-21-10-8-18/h4-7,20H,1-3,8-10,18H2/b19-13+
Formula Weight 304.1399
Percent Purity >95%
Chemical Name or Material Desmethyl Fluvoxamine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.