Learn More
Desmethyl Ofloxacin Hydrochloride, TRC
Product Code. 30134501
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134501 100 mg 100mg
30011686 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30134501 Supplier Toronto Research Chemicals Supplier No. TRCD292098100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 82419-52-1
Molecular Formula C17 H18 F N3 O4
Molecular Weight (g/mol) 347.34
Synonym 7H-Pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid, 9-fluoro-2,3-dihydro-3-methyl-7-oxo-10-(1-piperazinyl)-, 9-Fluoro-2,3-dihydro-3-methyl-7-oxo-10-(1-piperazinyl)-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid, (RS)-9-Fluoro-3-methyl-7-oxo-10-(piperazin-1-yl)-2,3-dihydro-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid, Desmethylofloxacin, Desmethyl Ofloxacin, Ofloxacin Imp. E (EP), Ofloxacin Rel. Comp. A (USP)
IUPAC Name (RS)-9-Fluoro-3-methyl-7-oxo-10-(piperazin-1-yl)-2,3-dihydro-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid
SMILES CC1COc2c(N3CCNCC3)c(F)cc4C(=O)C(=CN1c24)C(=O)O
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C17H18FN3O4/c1-9-8-25-16-13-10(15(22)11(17(23)24)7-21(9)13)6-12(18)14(16)20-4-2-19-3-5-20/h6-7,9,19H,2-5,8H2,1H3,(H,23,24)
Formula Weight 347.1281
Chemical Name or Material Desmethylofloxacin