Learn More
Desmethyl Ofloxacin, TRC
Product Code. 30137514
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137514 50 mg 50mg
30008603 10 mg 10mg
30010597 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30137514 Supplier Toronto Research Chemicals Supplier No. TRCD29227050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 82419-52-1
Molecular Formula C17 H18 F N3 O4
Molecular Weight (g/mol) 347.34
Synonym 7H-Pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid, 9-fluoro-2,3-dihydro-3-methyl-7-oxo-10-(1-piperazinyl)-, 9-Fluoro-2,3-dihydro-3-methyl-7-oxo-10-(1-piperazinyl)-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid, (RS)-9-Fluoro-3-methyl-7-oxo-10-(piperazin-1-yl)-2,3-dihydro-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid, Desmethylofloxacin, Desmethyl Ofloxacin, Ofloxacin Imp. E (EP), Ofloxacin Rel. Comp. A (USP)
IUPAC Name (RS)-9-Fluoro-3-methyl-7-oxo-10-(piperazin-1-yl)-2,3-dihydro-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid
SMILES CC1COc2c(N3CCNCC3)c(F)cc4C(=O)C(=CN1c24)C(=O)O
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C17H18FN3O4/c1-9-8-25-16-13-10(15(22)11(17(23)24)7-21(9)13)6-12(18)14(16)20-4-2-19-3-5-20/h6-7,9,19H,2-5,8H2,1H3,(H,23,24)
Formula Weight 347.1281
Percent Purity >95%
Chemical Name or Material Desmethylofloxacin
Grade HPLC