missing translation for 'onlineSavingsMsg'
Learn More

Desmethylfluvoxamine, Lipomed

Product Code. 30457728
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30457728 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30457728 Supplier Lipomed Supplier No. LPMFLV1889FB10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.

Desmethylfluvoxamine
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Desmethyl Fluvoxamine
CAS 192876-02-1
Analyte or Component Names Desmethyl Fluvoxamine
Shipping Condition Room Temperature
Molecular Formula C14 H19 F3 N2 O2
Synonym (1 E)-5-hydroxy-1-[4-(trifluoromethyl)phenyl]-1-Pentanone O-(2-aminoethyl)oxime, 1-Pentanone, 5-hydroxy-1-[4-(trifluoromethyl)phenyl]-, O-(2-aminoethyl)oxime, (E)-, Fluvoxamino acid, Fluvoxamine Maleate Imp. G (EP)
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES NCCO\N=C(/CCCCO)\c1ccc(cc1)C(F)(F)F
IUPAC Name (5 E)-5-(2-aminoethoxyimino)-5-[4-(trifluoromethyl)phenyl]pentan-1-ol
Industry Type Pharmaceutical
Molecular Weight (g/mol) 304.31
Quantity 10 mg
Formula Weight 304.1399 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.