Learn More
Desmethylofloxacin, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Desmethylofloxacin |
| CAS | 82419-52-1 |
| Analyte or Component Names | Desmethylofloxacin |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H18 F N3 O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Ofloxacin Imp. E (EP), Ofloxacin USP Related Compound A, Ofloxacin USP RC A, (RS)-9-Fluoro-3-methyl-7-oxo-10-(piperazin-1-yl)-2,3-dihydro-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic Acid, Ofloxacin Related Compound A, Ofloxacin Impurity E |
| InChI Formula | InChI=1S/C17H18FN3O4/c1-9-8-25-16-13-10(15(22)11(17(23)24)7-21(9)13)6-12(18)14(16)20-4-2-19-3-5-20/h6-7,9,19H,2-5,8H2,1H3,(H,23,24) |
| Solution Type | Neat |
| SMILES | CC1COc2c(N3CCNCC3)c(F)cc4C(=O)C(=CN1c24)C(=O)O |
| Show More |