Learn More
Desmin Antibody (CL4502) [DyLight 350], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-59056UV
Product Code. 30295128
  • € 737.00 / 0.10mL
Description
Desmin Monoclonal antibody specifically detects Desmin in Human samples. It is validated for Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Desmin
Monoclonal
DyLight 350
CMD1IFLJ41013, CSM1, CSM2, desmin, FLJ12025, FLJ39719, FLJ41793, intermediate filament protein
This antibody was developed using a synthetic peptide derived from P17661, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cell Biology, Cellular Markers, Phospho Specific
1674
Store at 4⪚C in the dark.
IgG2a
Western Blot, Immunocytochemistry/Immunofluorescence
CL4502
50mM Sodium Borate
Mouse
Protein A purified