missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Desmin Antibody (OTI4G1), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70565G
This item is not returnable.
View return policy
Description
Desmin Monoclonal specifically detects Desmin in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
| Desmin | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| DES | |
| Recombinant protein fragment corresponding to amino acids 104-391 of human Desmin (NP_001918) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| OTI4G1 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| CMD1IFLJ41013, CSM1, CSM2, desmin, FLJ12025, FLJ39719, FLJ41793, intermediate filament protein | |
| Mouse | |
| Affinity Purified | |
| Cytoskeleton Markers, Phospho Specific | |
| 1674.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction