missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Desmin Antibody (rDES/1711) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14038JF525
This item is not returnable.
View return policy
Description
Desmin Monoclonal antibody specifically detects Desmin in Chicken,Hamster,Human,Mouse,Rat samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Desmin | |
| Monoclonal | |
| Janelia Fluor 525 | |
| CMD1IFLJ41013, CSM1, CSM2, desmin, FLJ12025, FLJ39719, FLJ41793, intermediate filament protein | |
| Recombinant full-length human desmin protein (Uniprot: P17661) | |
| 0.1 mL | |
| Cell Biology, Cellular Markers, Phospho Specific | |
| 1674 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| rDES/1711 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Chicken, Hamster, Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction