Learn More
Anti-Human CD18 (TS1/18), Proteintech
Product Code. 30236737 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μg
500 μg
Unit Size:
100µg
500µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30236737 500 μg 500µg
30237865 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30236737 Supplier Proteintech Supplier No. 651901IG500UG
Mouse Monoclonal Antibody
The CD18 antibody from Proteintech is a mouse monoclonal antibody generated with human beta 2 integrin of human CD18. This antibody recognizes human antigen. The CD18 antibody has been validated for the following applications: FC analysis.
Specifications
Antigen CD18
Applications Flow Cytometry
Classification Monoclonal
Clone TS1/18
Concentration 500 μg/mL
Conjugate Unconjugated
Gene Symbols CD18; LAD; LCAMB; LFA-1; MAC-1; MF17; MFI7
Host Species Mouse
Immunogen Human Beta 2 Integrin
Purification Method Protein G purification
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3689
Target Species Human
Content And Storage Store at 2-8°C. Stable for one year after shipment.
Product Type Flow Antibody
Form Liquid
Isotype IgG1 κ