missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Desmocollin 3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583959
This item is not returnable.
View return policy
Description
Immunogen sequence: VTDSNDNAPT FRQNAYEAFV EENAFNVEIL RIPIEDKDLI NT Highest antigen sequence indentity to the following orthologs: Mouse - 79%, Rat - 76%.
Desmosomes are cell-cell junctions between epithelial, myocardial, and certain other cell types. Currently, three desmoglein subfamily members have been identified and all are members of the cadherin cell adhesion molecule superfamily. These desmoglein gene family members are located in a cluster on chromosome 18. This protein has been identified as the autoantigen of the autoimmune skin blistering disease pemphigus vulgaris.
Specifications
| Desmocollin 3 | |
| Polyclonal | |
| Unconjugated | |
| Dsc3 | |
| 5430426I24Rik; ARVC10; ARVD10; Cadherin family member 3; CDHF3; CDHF5; CDHF6; CMD1BB; desmocollin 3; desmocollin-3; Desmocollin-4; desmoglein-2; desmoglein-3; DSC; DSC1; DSC2; DSC3; DSC4; HDGC; HT-CP; PVA | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1825 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q14574 | |
| Dsc3 | |
| Recombinant protein corresponding to Human DSC3. Recombinant protein control fragment (Product #RP-100410). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction