missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Desmocollin 3 Recombinant Rabbit Monoclonal Antibody (JE55-86)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA542507
This item is not returnable.
View return policy
Description
Positive Control: Rat skin tissue lysate, A431 cell lysate, 293T, human skin tissue, mouse esophagus tissue, A431. Subcellular Location: Cell membrane, desmosome.
Desmosomes are cell-cell junctions between epithelial, myocardial, and certain other cell types. Currently, three desmoglein subfamily members have been identified and all are members of the cadherin cell adhesion molecule superfamily. These desmoglein gene family members are located in a cluster on chromosome 18. This protein has been identified as the autoantigen of the autoimmune skin blistering disease pemphigus vulgaris.
Specifications
| Desmocollin 3 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P55850, Q14574 | |
| Dsc3 | |
| Synthetic peptide within C-terminal Human Desmocollin 3. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Western Blot, Immunohistochemistry (Paraffin), Western Blot | |
| JE55-86 | |
| Unconjugated | |
| Dsc3 | |
| 5430426I24Rik; ARVC10; ARVD10; Cadherin family member 3; CDHF3; CDHF5; CDHF6; CMD1BB; desmocollin 3; desmocollin-3; Desmocollin-4; desmoglein-2; desmoglein-3; DSC; DSC1; DSC2; DSC3; DSC4; HDGC; HT-CP; PVA | |
| Rabbit | |
| Protein A | |
| RUO | |
| 13507, 1825, 307563 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto