missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Desmoglein-1 Mouse anti-Human, Clone: 32-2B, Novus Biologicals™
Mouse Monoclonal Antibody
€ 484.00
Specifications
| Antigen | Desmoglein-1 |
|---|---|
| Clone | 32-2B |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 1 to 2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
Description
Desmoglein-1 Monoclonal specifically detects Desmoglein-1 in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
| Desmoglein-1 | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human | |
| Cadherin family member 4, CDHF4PPKS1, desmoglein 1, desmoglein-1, Desmosomal glycoprotein 1, DG1, DGI, DSG, Pemphigus foliaceus antigen, SPPK1 | |
| Bovine desmoglein (desmosomal glyco protein I) from nasal epidermis. | |
| Primary | |
| Store at 4°C. |
| 32-2B | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 1828 | |
| IgG2a κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title