missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Desmoglein 1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106826
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total DSG1.
Desmosomes are cell-cell junctions between epithelial, myocardial, and certain other cell types. Currently, three desmoglein subfamily members have been identified and all are members of the cadherin cell adhesion molecule superfamily. These desmoglein gene family members are located in a cluster on chromosome 18. This protein has been identified as the autoantigen of the autoimmune skin blistering disease pemphigus vulgaris.
Specifications
| Desmoglein 1 | |
| Polyclonal | |
| Unconjugated | |
| DSG1 | |
| Cadherin family member 4; CDHF4; desmoglein 1; desmoglein 1 beta; desmoglein 1 gamma; desmoglein-1; Desmosomal glycoprotein 1; DG1; DSG; DSG1; Dsg1b; Dsg1c; EPKHE; EPKHIA; Pemphigus foliaceus antigen; PPKS1; SPPK1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 1828, 291755 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q02413 | |
| DSG1 | |
| A synthesized peptide derived from human DSG1(Accession Q02413), corresponding to amino acid residues A163-R213. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction