missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Desmoglein-2 Antibody ([8E5), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54529PE
This item is not returnable.
View return policy
Description
Desmoglein-2 Monoclonal antibody specifically detects Desmoglein-2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, CyTOF
Specifications
| Desmoglein-2 | |
| Monoclonal | |
| PE | |
| ARVC10, ARVD10, Cadherin family member 5, CDHF5MGC117034, CMD1BB, desmoglein 2, desmoglein-2, HDGC, MGC117036, MGC117037 | |
| Recombinant human Desmoglein-2 protein (Uniprot: Q14126) | |
| 100 μg | |
| Cancer, Cardiovascular Biology, Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 1829 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunofluorescence, CyTOF | |
| [8E5] | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction