missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Desmoglein-3 Antibody (6G2C11) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 196.00 - € 575.00
Specifications
| Antigen | Desmoglein-3 |
|---|---|
| Clone | 6G2C11 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18017615
|
Novus Biologicals
NBP2-37259 |
0.1 mL |
€ 575.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18625785
|
Novus Biologicals
NBP2-37259-0.025ml |
0.025 mL |
€ 196.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Desmoglein-3 Monoclonal specifically detects Desmoglein-3 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
| Desmoglein-3 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
| Unconjugated | |
| RUO | |
| Human | |
| Cadherin family member 6, CDHF6130-kD pemphigus vulgaris antigen, desmoglein 3, desmoglein 3 (pemphigus vulgaris antigen), desmoglein-3, DKFZp686P23184,130 kDa pemphigus vulgaris antigen, pemphigus vulgaris antigen, PVA | |
| DSG3 | |
| IgG1 | |
| Affinity Purified |
| 6G2C11 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Cytoskeleton Markers, Extracellular Matrix, Immune System Diseases, Signal Transduction | |
| PBS with 0.05% Sodium Azide | |
| 1830 | |
| Purified recombinant fragment of human DSG3 (AA: 55-159) expressed in E. Coli. | |
| Primary | |
| 107.5 kDa |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts