missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Desmoglein-3 Antibody (DSG3/1535), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08409AF700
This item is not returnable.
View return policy
Description
Desmoglein-3 Monoclonal antibody specifically detects Desmoglein-3 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications
| Desmoglein-3 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| Cadherin family member 6, CDHF6130-kD pemphigus vulgaris antigen, desmoglein 3, desmoglein 3 (pemphigus vulgaris antigen), desmoglein-3, DKFZp686P23184,130 kDa pemphigus vulgaris antigen, pemphigus vulgaris antigen, PVA | |
| Recombinant human Desmoglein-3 extracellular protein fragment (Uniprot: P32926) | |
| 100 μg | |
| Cell Biology, Cellular Markers, Extracellular Matrix, Immune System Diseases, Signal Transduction | |
| 1830 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Immunohistochemistry (Paraffin) | |
| DSG3/1535 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction