Learn More
Desmoglein-3 Antibody (rDSG3/8611), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals (Bio-Techne) NBP3-24060CL1
Product Code. 30150279
  • € 1075.00 / 0.10mL
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
Desmoglein-3 Monoclonal antibody specifically detects Desmoglein-3 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Spécification
Spécification
Desmoglein-3
Monoclonal
CoraFluor 1
Cadherin family member 6, CDHF6130-kD pemphigus vulgaris antigen, desmoglein 3, desmoglein 3 (pemphigus vulgaris antigen), desmoglein-3, DKFZp686P23184,130 kDa pemphigus vulgaris antigen, pemphigus vulgaris antigen, PVA
Recombinant fragment (around aa300-500) of human Desmoglein-3 protein (exact sequence is proprietary)
0.1 mL
Cell Biology, Cellular Markers, Extracellular Matrix, Immune System Diseases, Signal Transduction
1830
Store at 4°C in the dark. Do not freeze.
IgG1 κ
Immunohistochemistry (Paraffin)
rDSG3/8611