missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Desmoplakin Monoclonal Antibody (DP-2.15)
GREENER_CHOICE

Product Code. 11534603
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11534603 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11534603 Supplier Invitrogen™ Supplier No. MA183118

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Mouse anti Desmoplakin 1/2 antibody, clone DP-2.15 recognizes both desmoplakin 1 and 2 from stratified epithelia, simple epithelia including glands, urothelium, thymic reticular epithelium, hepatocytes, intercalated disks of myocardium and arachnoid cells of meninges. Mouse anti Desmoplakin 1/2 antibody, clone DP-2.15 can be used for the detection of primary and metastatic carcinomas.

Desmoplakin (DSP) is a major high molecular weight protein of the desmosomes. It is involved in the organization of the desmosomal cadherin-plakoglobin complexes into discrete plasma membrane domains and in the anchoring of intermediate filaments to the desmosomes. Desmoplakin mutations are associated with Keratoderma and cardiomyopathy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Desmoplakin
Applications Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone DP-2.15
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with <0.1% sodium azide; pH 7.4
Gene DSP
Gene Accession No. E9Q557, P15924
Gene Alias 2300002E22Rik; 250/210 kDa paraneoplastic pemphigus antigen; 5730453H04Rik; AA407887; AA407888; AW109828; DCWHKTA; dermatan sulfate proteoglycan 3; desmoplakin; desmoplakin I/II; Desmoplakin II; desmoplakin; LOW QUALITY PROTEIN: desmoplakin; Desmoplakin-I; desmosomal cytoskeletal connector molecule; DP; Dsp; dspg3; Oculoglycan; Opt; Optc; optcl; Opticin; rul; SLRP; SLRP protein; small leucine-rich repeat protein; small leucine-rich repeat protein (SLRP); wu:fb81a05; zgc:101047
Gene Symbols DSP
Host Species Mouse
Immunogen Native bovine desmoplakin 1 and 2.
Purification Method Protein A
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 109620, 1832, 306871, 420869, 514360
Target Species Human, Mouse, Rat, Bovine, Chicken
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.