missing translation for 'onlineSavingsMsg'
Learn More

Desoxypipradrol, TRC

Product Code. 30367643
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30367643 1 g 1g
30363135 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30367643 Supplier Toronto Research Chemicals Supplier No. TRCD2969801G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 519-74-4
Molecular Formula C8H21N
Molecular Weight (g/mol) 251.37
Synonym 2-(Diphenylmethyl)piperidine, (±)-Desoxypipradrol, 2-Diphenylmethylpiperidine, Ciba 14469, Deoxypipradrol, Desoxypipradrol
SMILES C1CCC(NC1)C(c2ccccc2)c3ccccc3

Specifications

Quantity 1 g
InChI Formula InChI=1S/C18H21N/c1-3-9-15(10-4-1)18(16-11-5-2-6-12-16)17-13-7-8-14-19-17/h1-6,9-12,17-19H,7-8,13-14H2
Formula Weight 251.1674
Chemical Name or Material Desoxypipradrol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.